2-(4-Bromobenzyl)-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR17895-1G
APOSOR17895-1GEA 0 EUR
APOSOR17895-1G
2-(4-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
2-(4-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
Formel: C₁₁H₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD09258846
CAS-Nummer: 1086380-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR