2-(3-Methoxybenzyl)-1,3-thiazole-4-carboxylic acid

Lieferant: Apollo Scientific

OR17896-1G
APOSOR17896-1GEA 0 EUR
APOSOR17896-1G
2-(3-Methoxybenzyl)-1,3-thiazole-4-carboxylic acid
2-(3-Methoxybenzyl)-1,3-thiazole-4-carboxylic acid
Formel: C₁₂H₁₁NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD09258848
CAS-Nummer: 1086380-17-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR