2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
Lieferant: Apollo Scientific
OR17897-1G
APOSOR17897-1GEA
0
EUR
APOSOR17897-1G
2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
2-(2-Bromobenzyl)-1,3-thiazole-4-carboxylic acid
Formel:
C₁₁H₈BrNO₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258847 CAS-Nummer: 1086380-14-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...