2-(4-Hydroxyphenyl)propionsäure
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(4-Hydroxyphenyl)propionic Acid
OR17909-10G
OR17909-1G
OR17909-50G
APOSOR17909-10GEA
121
EUR
APOSOR17909-10G
APOSOR17909-1G
APOSOR17909-50G
2-(4-Hydroxyphenyl)propionsäure
2-(4-Hydroxyphenyl)propionsäure
Formel:
C₉H₁₀O₃ Schmelzpunkt: 130…132 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128847 CAS-Nummer: 938-96-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...