2-Amino-5-hydroxy-1,3-benzothiazole

Lieferant: Apollo Scientific
Achtung

OR17933-250MG
APOSOR17933-250MGEA 0 EUR
APOSOR17933-250MG
2-Amino-5-hydroxy-1,3-benzothiazole
2-Amino-5-hydroxy-1,3-benzothiazole
Formel: C₇H₆N₂OS
Lagertemperatur: Raumtemperatur
MDL: MFCD00985125
CAS-Nummer: 118526-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR