2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98+%

Lieferant: Apollo Scientific
Achtung

OR17939-500G
APOSOR17939-500GEA 109 EUR
APOSOR17939-500G
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione 98+%
2-(Methylthio)pyrimidine-4,6(1H,5H)-dione
Formel: C₅H₆N₂O₂S
Lagertemperatur: Kühlschrank
MDL: MFCD00810414
CAS-Nummer: 1979-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR