2-Amino-4,6-dichloropyrimidine-5-carbonitrile 95+%

Lieferant: Apollo Scientific

OR17955-1G
APOSOR17955-1GEA 40.6 EUR
APOSOR17955-1G
2-Amino-4,6-dichloropyrimidine-5-carbonitrile 95+%
2-Amino-4,6-dichloropyrimidine-5-carbonitrile
Formel: C₅H₂Cl₂N₄
Molecular Weight: 189,002 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20727436
CAS-Nummer: 1277179-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR