tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-{2-[(tert-Butoxycarbonyl)amino]ethoxy}aniline, tert-Butyl 2-(4-aminophenoxy)ethylcarbamate

OR17990-250MG
APOSOR17990-250MGEA 24.8 EUR
APOSOR17990-250MG
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate
tert-Butyl [2-(4-aminophenoxy)ethyl]carbamate
Formel: C₁₃H₂₀N₂O₃
Molecular Weight: 252,309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11922406
CAS-Nummer: 159184-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR