2-(1,3-Benzoxazol-2-yl)-1-(4-tert-butylphenyl)ethanone 95+%

Lieferant: Apollo Scientific
Achtung

OR18001-1G
APOSOR18001-1GEA 98.6 EUR
APOSOR18001-1G
2-(1,3-Benzoxazol-2-yl)-1-(4-tert-butylphenyl)ethanone 95+%
2-(1,3-Benzoxazol-2-yl)-1-(4-tert-butylphenyl)ethanone
Formel: C₁₉H₁₉NO₂
Molecular Weight: 293,37 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04037909
CAS-Nummer: 849021-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR