8-Amino-4-methylchinolin
Lieferant: Apollo Scientific
|
Achtung
|
OR18011-250MG
OR18011-500MG
APOSOR18011-250MGEA
120
EUR
APOSOR18011-250MG
APOSOR18011-500MG
8-Amino-4-methylchinolin
8-Amino-4-methylquinoline
Formel:
C₁₀H₁₀N₂ Lagertemperatur: Kühlschrank |
MDL:
MFCD03819702 CAS-Nummer: 62748-01-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...