3-Oxoindane-4-carbonitrile 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-3-oxo-1H-indene-4-carbonitrile, 4-Cyano-3-oxoindane

OR18013-250MG
APOSOR18013-250MGEA 235 EUR
APOSOR18013-250MG
3-Oxoindane-4-carbonitrile 95%
3-Oxoindane-4-carbonitrile
Formel: C₁₀H₇NO
Molecular Weight: 157,169 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11035917
CAS-Nummer: 215362-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR