3-Methyl-2-nitrophenylacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Carboxymethyl)-6-methylnitrobenzene, 3-(Carboxymethyl)-2-nitrotoluene, 2-(3-Methyl-2-nitrophenyl)acetic acid

OR18016-250MG
APOSOR18016-250MGEA 79.9 EUR
APOSOR18016-250MG
3-Methyl-2-nitrophenylacetic acid
3-Methyl-2-nitrophenylacetic acid
Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08276006
CAS-Nummer: 18710-86-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR