6-Amino-3-bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-Bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine

OR18021-1G
APOSOR18021-1GEA 203 EUR
APOSOR18021-1G
6-Amino-3-bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidine
6-Amino-3-bromo-4-chloro-1H-pyrazolo[3,4-d]pyrimidine
Formel: C₅H₃BrClN₅
Molecular Weight: 248,468 g/mol
Schmelzpunkt: 267…273 °C
Lagertemperatur: Kühlschrank
MDL: MFCD13193632
CAS-Nummer: 1003298-79-0
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR