2,5-Dibromthieno[3,2-b]thiophen 98%
Lieferant: Apollo Scientific
|
Gefahr
|
OR18024-5G
APOSOR18024-5GEA
145
EUR
APOSOR18024-5G
2,5-Dibromthieno[3,2-b]thiophen 98%
2,5-Dibromothieno[3,2-b]thiophene
Formel:
C₆H₂Br₂S₂ Molecular Weight: 298.0181 g/mol Schmelzpunkt: 124…129 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD03931293 CAS-Nummer: 25121-87-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...