6-Chinoxalincarbonsäurepinakolester 95+%

Lieferant: Apollo Scientific

Synonyme: Quinoxaline-6-boronic acid, pinacol ester, 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)quinoxaline

OR18030-1G
APOSOR18030-1GEA 155 EUR
APOSOR18030-1G
6-Chinoxalincarbonsäurepinakolester 95+%
6-Chinoxalincarbonsäurepinakolester
Formel: C₁₄H₁₇BN₂O₂
Molecular Weight: 256.1080 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11054040
CAS-Nummer: 1167418-13-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR