N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-serin
Lieferant: Apollo Scientific
Synonyme:
(S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate
OR18040-10G
OR18040-250G
OR18040-50G
APOSOR18040-10GEA
21.9
EUR
APOSOR18040-10G
APOSOR18040-250G
APOSOR18040-50G
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-serin
N-Boc-L-serine
Formel:
C₈H₁₅NO₅ Schmelzpunkt: 91 °C Flammpunkt: 186 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037243 CAS-Nummer: 3262-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...