N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-serin

Lieferant: Apollo Scientific

Synonyme: (S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxypropanoic acid, Boc-Ser-OH, N-(tert-Butoxycarbonyl)-L-serine, Boc-L-serine, N-(tert-Butoxycarbonyl)-L-serine hydrate, BOC-L-Serine hydrate

OR18040-10G OR18040-250G OR18040-50G
APOSOR18040-10GEA 21.9 EUR
APOSOR18040-10G APOSOR18040-250G APOSOR18040-50G
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-L-serin
N-Boc-L-serine
Formel: C₈H₁₅NO₅
Schmelzpunkt: 91 °C
Flammpunkt: 186 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00037243
CAS-Nummer: 3262-72-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR