5-Chloro-2-ethoxy-nicotinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chloro-2-ethoxynicotinic acid

OR18046-1G OR18046-250MG OR18046-5G
APOSOR18046-1GEA 31.9 EUR
APOSOR18046-1G APOSOR18046-250MG APOSOR18046-5G
5-Chloro-2-ethoxy-nicotinic acid
5-Chloro-2-ethoxy-nicotinic acid
Formel: C₈H₈ClNO₃
Molecular Weight: 201.61 g/mol
Schmelzpunkt: 115.5…117.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07403147
CAS-Nummer: 68359-07-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR