2-Chloro-N-methylpyrimidin-4-amine

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-4-(methylamino)pyrimidine

OR18111-1G
APOSOR18111-1GEA 59.6 EUR
APOSOR18111-1G
2-Chloro-N-methylpyrimidin-4-amine
2-Chloro-N-methylpyrimidin-4-amine
Formel: C₅H₆ClN₃
Molecular Weight: 143,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09055368
CAS-Nummer: 66131-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR