5-Amino-4-chloro-6-methoxypyrimidine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-6-methoxypyrimidin-5-amine
OR18113-1G
APOSOR18113-1GEA
119
EUR
APOSOR18113-1G
5-Amino-4-chloro-6-methoxypyrimidine
5-Amino-4-chloro-6-methoxypyrimidine
Formel:
C₅H₆ClN₃O Molecular Weight: 159,574 g/mol Schmelzpunkt: 70…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646119 CAS-Nummer: 15846-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...