6-Iodopyridazin-3-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Amino-6-iodopyridazine

OR18114-1G
APOSOR18114-1GEA 24.8 EUR
APOSOR18114-1G
6-Iodopyridazin-3-amine
6-Iodopyridazin-3-amine
Formel: C₄H₄IN₃
Molecular Weight: 221 g/mol
Schmelzpunkt: 158…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09991754
CAS-Nummer: 187973-60-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR