Sodium-1-naphthalenesulphonate 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR18282-100G
OR18282-25G
OR18282-500G
APOSOR18282-100GEA
0
EUR
APOSOR18282-100G
APOSOR18282-25G
APOSOR18282-500G
Sodium-1-naphthalenesulphonate 98%
Sodium-1-naphthalenesulphonate
Formel:
C₁₀H₈O₃S Schmelzpunkt: 299…301 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064964 CAS-Nummer: 130-14-3 EINECS: 204-976-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...