2-Amino-2',5-dichlorbenzophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2-Amino-5-chlorophenyl)(2-chlorophenyl)methanone
OR18295-5G
APOSOR18295-5GEA
21.9
EUR
APOSOR18295-5G
2-Amino-2',5-dichlorbenzophenon 98%
2-Amino-2',5-dichlorbenzophenon
Formel:
C₁₃H₉Cl₂NO Molecular Weight: 266,13 g/mol Schmelzpunkt: 87…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007840 CAS-Nummer: 2958-36-3 EINECS: 220-985-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...