[2.2]-Paracyclophan 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR18296-25G
OR18296-5G
OR18296-100G
APOSOR18296-25GEA
40.6
EUR
APOSOR18296-25G
APOSOR18296-5G
APOSOR18296-100G
[2.2]-Paracyclophan 98%
[2.2]-Paracyclophan
Formel:
C₁₆H₁₆ Schmelzpunkt: 283…288 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003707 CAS-Nummer: 1633-22-3 EINECS: 216-644-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...