O-Methylhydroxylamin Hydrochlorid 98%

Lieferant: Apollo Scientific
Gefahr

OR18305-100G OR18305-25G OR18305-500G
APOSOR18305-100GEA 0 EUR
APOSOR18305-100G APOSOR18305-25G APOSOR18305-500G
O-Methylhydroxylamin Hydrochlorid 98%
O-Methylhydroxylamin Hydrochlorid
Formel: CH₆ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD00012951
CAS-Nummer: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR