6-Bromo-5-chloronicotinic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-5-chloropyridine-3-carboxylic acid

OR183414-250MG
APOSOR183414-250MGEA 473 EUR
APOSOR183414-250MG
6-Bromo-5-chloronicotinic acid 95%
6-Bromo-5-chloronicotinic acid
Formel: C₆H₃BrClNO₂
Molecular Weight: 236,45 g/mol
Schmelzpunkt: 158…163 °C
MDL: MFCD03426922
CAS-Nummer: 38185-54-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR