5-Bromo-6-chloropicolinic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-6-chloropyridine-2-carboxylic acid

OR183415-250MG OR183415-1G OR183415-100MG
APOSOR183415-250MGEA 109 EUR
APOSOR183415-250MG APOSOR183415-1G APOSOR183415-100MG
5-Bromo-6-chloropicolinic acid
5-Bromo-6-chloropicolinic acid
Formel: C₆H₃BrClNO₂
Molecular Weight: 236.451 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09910220
CAS-Nummer: 959958-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR