Ethyl 3-amino-5-nitrobenzo[b]thiophene-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR183422-1G OR183422-250MG
APOSOR183422-1GEA 261 EUR
APOSOR183422-1G APOSOR183422-250MG
Ethyl 3-amino-5-nitrobenzo[b]thiophene-2-carboxylate
Ethyl 3-amino-5-nitrobenzo[b]thiophene-2-carboxylate
Formel: C₁₁H₁₀N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD20922023
CAS-Nummer: 27697-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR