3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR183423-1G OR183423-250MG
APOSOR183423-1GEA 364 EUR
APOSOR183423-1G APOSOR183423-250MG
3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid
3-Amino-5-nitrobenzo[b]thiophene-2-carboxylic acid
Formel: C₉H₆N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD20924150
CAS-Nummer: 160041-48-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR