Methyl-3-amino-4-chlor-2-benzo[b]thiophencarboxylat

Lieferant: Apollo Scientific

OR183424-1G
APOSOR183424-1GEA 116 EUR
APOSOR183424-1G
Methyl-3-amino-4-chlor-2-benzo[b]thiophencarboxylat
Methyl-3-amino-4-chlor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₈ClNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01566406
CAS-Nummer: 35212-86-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR