Methyl 3-amino-7-iodobenzothiophene-2-carboxylate

Lieferant: Apollo Scientific

OR183430-250MG
APOSOR183430-250MGEA 335 EUR
APOSOR183430-250MG
Methyl 3-amino-7-iodobenzothiophene-2-carboxylate
Methyl 3-amino-7-iodobenzothiophene-2-carboxylate
Formel: C₁₀H₈INO₂S
Molecular Weight: 333,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29163661
CAS-Nummer: 2089797-56-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR