2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR183431-1G
APOSOR183431-1GEA 516 EUR
APOSOR183431-1G
2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2-Methyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Formel: C₁₁H₁₀N₄
Molecular Weight: 198,224 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269358
CAS-Nummer: 264226-27-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR