2,6-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR183432-1G
APOSOR183432-1GEA 219 EUR
APOSOR183432-1G
2,6-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2,6-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Formel: C₁₂H₁₂N₄
Molecular Weight: 212,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269369
CAS-Nummer: 372198-37-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR