2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR183433-1G
APOSOR183433-1GEA 219 EUR
APOSOR183433-1G
2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2,7-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Formel: C₁₂H₁₂N₄
Molecular Weight: 212.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00269378
CAS-Nummer: 372198-31-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR