3-(1H-Pyrazol-3-yl)-1H-indole
Lieferant: Apollo Scientific
OR183434-1G
APOSOR183434-1GEA
219
EUR
APOSOR183434-1G
3-(1H-Pyrazol-3-yl)-1H-indole
3-(1H-Pyrazol-3-yl)-1H-indole
Formel:
C₁₁H₉N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06808799 CAS-Nummer: 29903-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...