3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine

Lieferant: Apollo Scientific

OR183435-1G
APOSOR183435-1GEA 219 EUR
APOSOR183435-1G
3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine
3-[4-Methyl-5-(1H-pyrazol-3-yl)-1,3-thiazol-2-yl]pyridine
Formel: C₁₂H₁₀N₄S
Molecular Weight: 242,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06808800
CAS-Nummer: 499796-05-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR