3-(1H-Pyrrol-2-yl)-1H-pyrazole

Lieferant: Apollo Scientific

OR183437-1G
APOSOR183437-1GEA 219 EUR
APOSOR183437-1G
3-(1H-Pyrrol-2-yl)-1H-pyrazole
3-(1H-Pyrrol-2-yl)-1H-pyrazole
Formel: C₇H₇N₃
Molecular Weight: 133,151 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06808816
CAS-Nummer: 1019010-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR