N,N-Dimethyl-4-(1H-pyrazol-3-yl)benzenesulphonamide

Lieferant: Apollo Scientific

OR183438-1G
APOSOR183438-1GEA 219 EUR
APOSOR183438-1G
N,N-Dimethyl-4-(1H-pyrazol-3-yl)benzenesulphonamide
N,N-Dimethyl-4-(1H-pyrazol-3-yl)benzenesulphonamide
Formel: C₁₁H₁₃N₃O₂S
Molecular Weight: 251.305 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 1019011-14-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR