4-[4-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine

Lieferant: Apollo Scientific

OR183440-1G
APOSOR183440-1GEA 219 EUR
APOSOR183440-1G
4-[4-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine
4-[4-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine
Formel: C₁₃H₁₅N₃O₃S
Molecular Weight: 293,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774261
CAS-Nummer: 1019011-88-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR