N,N-Dimethyl-3-(1H-pyrazol-3-yl)benzenesulphonamide
Lieferant: Apollo Scientific
OR183442-1G
APOSOR183442-1GEA
219
EUR
APOSOR183442-1G
N,N-Dimethyl-3-(1H-pyrazol-3-yl)benzenesulphonamide
N,N-Dimethyl-3-(1H-pyrazol-3-yl)benzenesulphonamide
Formel:
C₁₁H₁₃N₃O₂S Molecular Weight: 251,305 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07774271 CAS-Nummer: 1019012-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...