4-[3-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine

Lieferant: Apollo Scientific

OR183444-1G
APOSOR183444-1GEA 219 EUR
APOSOR183444-1G
4-[3-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine
4-[3-(1H-Pyrazol-3-yl)phenyl]sulphonylmorpholine
Formel: C₁₃H₁₅N₃O₃S
Molecular Weight: 293,342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07774281
CAS-Nummer: 1019013-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR