1-Methyl-4-[3-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine

Lieferant: Apollo Scientific

OR183445-1G
APOSOR183445-1GEA 0 EUR
APOSOR183445-1G
1-Methyl-4-[3-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine
1-Methyl-4-[3-(1H-pyrazol-3-yl)phenyl]sulphonyl-piperazine
Formel: C₁₄H₁₈N₄O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD07774286
CAS-Nummer: 1019013-80-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR