2,8-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine

Lieferant: Apollo Scientific

OR183447-1G
APOSOR183447-1GEA 219 EUR
APOSOR183447-1G
2,8-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
2,8-Dimethyl-3-(1H-pyrazol-3-yl)imidazo[1,2-a]pyridine
Formel: C₁₂H₁₂N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00269634
CAS-Nummer: 372198-36-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR