3-(1H-Pyrazol-3-yl)benzamide
Lieferant: Apollo Scientific
OR183448-1G
APOSOR183448-1GEA
219
EUR
APOSOR183448-1G
3-(1H-Pyrazol-3-yl)benzamide
3-(1H-Pyrazol-3-yl)benzamide
Formel:
C₁₀H₉N₃O Molecular Weight: 187,198 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08059381 CAS-Nummer: 1019015-64-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...