3-Phenyl-5-(1H-pyrazol-3-yl)isoxazole

Lieferant: Apollo Scientific

OR183449-1G
APOSOR183449-1GEA 219 EUR
APOSOR183449-1G
3-Phenyl-5-(1H-pyrazol-3-yl)isoxazole
3-Phenyl-5-(1H-pyrazol-3-yl)isoxazole
Formel: C₁₂H₉N₃O
Molecular Weight: 211,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00832976
CAS-Nummer: 264225-67-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR