3-(2,5-Dimethoxyphenyl)-1H-pyrazol

Lieferant: Apollo Scientific

OR183450-1G
APOSOR183450-1GEA 145 EUR
APOSOR183450-1G
3-(2,5-Dimethoxyphenyl)-1H-pyrazol
3-(2,5-Dimethoxyphenyl)-1H-pyrazole
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091528
CAS-Nummer: 181122-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR