3-(2,4-Dimethoxyphenyl)-1H-pyrazol

Lieferant: Apollo Scientific

OR183451-1G
APOSOR183451-1GEA 145 EUR
APOSOR183451-1G
3-(2,4-Dimethoxyphenyl)-1H-pyrazol
3-(2,4-Dimethoxyphenyl)-1H-pyrazole
Formel: C₁₁H₁₂N₂O₂
Molecular Weight: 204,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02091529
CAS-Nummer: 150433-17-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR