2-(1H-Pyrazol-5-yl)anilin
Lieferant: Apollo Scientific
Synonyme:
3-(2-Aminophenyl)-1H-pyrazole, 2-(1H-Pyrazol-3-yl)aniline
OR183453-250MG
APOSOR183453-250MGEA
58
EUR
APOSOR183453-250MG
2-(1H-Pyrazol-5-yl)anilin
2-(1H-Pyrazol-5-yl)anilin
Formel:
C₉H₉N₃ Molecular Weight: 159.1879 g/mol Schmelzpunkt: 129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028476 CAS-Nummer: 111562-32-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...