N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
Lieferant: Apollo Scientific
OR183454-1G
APOSOR183454-1GEA
183
EUR
APOSOR183454-1G
N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
N-[3-(1H-Pyrazol-3-yl)phenyl]acetamide
Formel:
C₁₁H₁₁N₃O Molecular Weight: 201,225 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09028477 CAS-Nummer: 197093-24-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...