3-[1,1'-Biphenyl]-4-yl-1H-pyrazole

Lieferant: Apollo Scientific

OR183455-1G
APOSOR183455-1GEA 145 EUR
APOSOR183455-1G
3-[1,1'-Biphenyl]-4-yl-1H-pyrazole
3-[1,1'-Biphenyl]-4-yl-1H-pyrazole
Formel: C₁₅H₁₂N₂
Molecular Weight: 220,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02755978
CAS-Nummer: 446276-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR